Learn More
Meso Scale Discovery U-PLEX NHP IL-23 ASY 25 PL
SDP
Catalog No. 5000080067
Encompass
The Meso Scale Discovery U-PLEX NHP IL-23 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human and non-human primate interleukin 23 (IL-23) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 1 3 pg/mL This assay is for research use only and not for use in diagnostic procedures It is available in a kit format that includes a 96-well plate coated with capture antibody detection antibody read buffer 10X diluent and calibrator
Catalog No. 50-000-80067 Supplier Meso Scale Discovery Supplier No. K156WGK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-23 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human and non-human primate interleukin 23 (IL-23) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 1 3 pg/mL This assay is for research use only and not for use in diagnostic procedures It is available in a kit format that includes a 96-well plate coated with capture antibody detection antibody read buffer 10X diluent and calibrator