Learn More
Meso Scale Discovery U-PLEX NHP IL-23 ASY
SDP
Catalog No. 5000079777
Encompass
The Meso Scale Discovery U-PLEX NHP IL-23 Assay is a multiplex immunoassay kit for measuring human and non-human primate IL-23 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls Turnaround time is under 3 5 hours for up to 80 samples aiding in immune response and inflammation studies
Catalog No. 50-000-79777 Supplier Meso Scale Discovery Supplier No. K156WGK2
The Meso Scale Discovery U-PLEX NHP IL-23 Assay is a multiplex immunoassay kit for measuring human and non-human primate IL-23 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls Turnaround time is under 3 5 hours for up to 80 samples aiding in immune response and inflammation studies