Learn More
Meso Scale Discovery U-PLEX NHP IL-23 ASY
SDP
Catalog No. 5000080714
Encompass
The Meso Scale Discovery U-PLEX NHP IL-23 Assay is a multiplex immunoassay kit for measuring human and non-human primate IL-23 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies for IL-23p19 and IL-12p40 detection antibodies with MSD SULFO-TAG labels and calibrators for a standard curve enabling sensitive reproducible and rapid IL-23 quantification
Catalog No. 50-000-80714 Supplier Meso Scale Discovery Supplier No. K156WGK1
The Meso Scale Discovery U-PLEX NHP IL-23 Assay is a multiplex immunoassay kit for measuring human and non-human primate IL-23 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies for IL-23p19 and IL-12p40 detection antibodies with MSD SULFO-TAG labels and calibrators for a standard curve enabling sensitive reproducible and rapid IL-23 quantification