Learn More
Meso Scale Discovery U-PLEX NHP IL-23 ASY384-well
SDP
Catalog No. 5000080266
Encompass
The Meso Scale Discovery U-PLEX NHP IL-23 384-Well Assay Kit is designed for quantifying IL-23 in human cynomolgus monkey and mouse samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 384-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators and buffers It has a sensitivity of 0 3 pg/mL and an upper limit of 10 000 pg/mL
Catalog No. 50-000-80266 Supplier Meso Scale Discovery Supplier No. K256WGK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-23 384-Well Assay Kit is designed for quantifying IL-23 in human cynomolgus monkey and mouse samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results with low sample volume The kit includes a 384-well plate with capture antibodies detection antibodies with an electrochemiluminescent label calibrators and buffers It has a sensitivity of 0 3 pg/mL and an upper limit of 10 000 pg/mL