Learn More
Meso Scale Discovery U-PLEX NHP IL-23 ASYQP 25 PL
SDP
Catalog No. 5000080066
Encompass
The U-PLEX NHP IL-23 Assay (K156WGK) is a multiplex immunoassay designed for quantitative measurement of interleukin-23 in non-human primate samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity and specificity for cytokine detection in research applications The kit provides reliable reproducible results for studying inflammatory responses and immune system function in preclinical studies using primate models
Catalog No. 50-000-80066 Supplier Meso Scale Discovery Supplier No. K156WGK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP IL-23 Assay (K156WGK) is a multiplex immunoassay designed for quantitative measurement of interleukin-23 in non-human primate samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity and specificity for cytokine detection in research applications The kit provides reliable reproducible results for studying inflammatory responses and immune system function in preclinical studies using primate models