Learn More
Meso Scale Discovery U-PLEX NHP IL-23 ASYQP
SDP
Catalog No. 5000080676
Encompass
The U-PLEX NHP IL-23 Assay (K156WGK) is a multiplex immunoassay designed for quantitative measurement of interleukin-23 in non-human primate samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity and specificity for cytokine detection in research applications The kit provides reagents and protocols optimized for serum plasma and cell culture supernatant sample types enabling researchers to study inflammatory responses and immune system function in preclinical studies
Catalog No. 50-000-80676 Supplier Meso Scale Discovery Supplier No. K156WGK22
The U-PLEX NHP IL-23 Assay (K156WGK) is a multiplex immunoassay designed for quantitative measurement of interleukin-23 in non-human primate samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity and specificity for cytokine detection in research applications The kit provides reagents and protocols optimized for serum plasma and cell culture supernatant sample types enabling researchers to study inflammatory responses and immune system function in preclinical studies