Learn More
Meso Scale Discovery U-PLEX NHP IL-4 ASY
SDP
Catalog No. 5000079969
Encompass
The Meso Scale Discovery U-PLEX NHP IL-4 Assay Kit is a sensitive immunoassay for quantifying natural human IL-4 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and a low detection limit Key features include a detection range of 0 05-1000 pg/mL a sample volume of 25 L a 3 5-hour assay time and no interference from IL-4 receptor This advanced kit is ideal for accurate IL-4 quantification in human samples
Catalog No. 50-000-79969 Supplier Meso Scale Discovery Supplier No. K156TWK2
The Meso Scale Discovery U-PLEX NHP IL-4 Assay Kit is a sensitive immunoassay for quantifying natural human IL-4 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and a low detection limit Key features include a detection range of 0 05-1000 pg/mL a sample volume of 25 L a 3 5-hour assay time and no interference from IL-4 receptor This advanced kit is ideal for accurate IL-4 quantification in human samples