Learn More
Meso Scale Discovery U-PLEX NHP IL-4 ASY
SDP
Catalog No. 5000081394
Encompass
The Meso Scale Discovery U-PLEX NHP IL-4 Assay Kit is a sensitive immunoassay for measuring natural human IL-4 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and a low detection limit of 0 03 pg/mL Key features include minimal matrix interference and high-throughput capabilities with results in 3 5 hours The kit includes a 96-well plate with capture antibody detection antibody read buffer wash buffer diluent calibrator and positive control
Catalog No. 50-000-81394 Supplier Meso Scale Discovery Supplier No. K156TWK1
The Meso Scale Discovery U-PLEX NHP IL-4 Assay Kit is a sensitive immunoassay for measuring natural human IL-4 in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a wide dynamic range and a low detection limit of 0 03 pg/mL Key features include minimal matrix interference and high-throughput capabilities with results in 3 5 hours The kit includes a 96-well plate with capture antibody detection antibody read buffer wash buffer diluent calibrator and positive control