Learn More
Meso Scale Discovery U-PLEX NHP IL-4 ASY384-well
SDP
Catalog No. 5000081231
Encompass
The Meso Scale Discovery U-PLEX NHP IL-4 384-Well Assay Kit is an electrochemiluminescent immunoassay for quantifying IL-4 in human non-human primate rat and mouse samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible IL-4 measurement with minimal interference The kit includes a 384-well plate with IL-4 capture antibody detection antibody calibrator diluent buffers and a lyophilized calibrator Sensitivity is 0 3 pg/mL with an upper quantitation limit of 1500 pg/mL
Catalog No. 50-000-81231 Supplier Meso Scale Discovery Supplier No. K256TWK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-4 384-Well Assay Kit is an electrochemiluminescent immunoassay for quantifying IL-4 in human non-human primate rat and mouse samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible IL-4 measurement with minimal interference The kit includes a 384-well plate with IL-4 capture antibody detection antibody calibrator diluent buffers and a lyophilized calibrator Sensitivity is 0 3 pg/mL with an upper quantitation limit of 1500 pg/mL