Learn More
Meso Scale Discovery U-PLEX NHP IL-4 ASYQP
SDP
Catalog No. 5000079915
Encompass
The U-PLEX NHP IL-4 Assay (K156TWK) is a specialized immunoassay kit designed for quantitative measurement of interleukin-4 (IL-4) in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for cytokine detection in research applications The kit provides researchers with validated reagents and protocols for studying immune responses and inflammatory processes in preclinical primate models
Catalog No. 50-000-79915 Supplier Meso Scale Discovery Supplier No. K156TWK22
The U-PLEX NHP IL-4 Assay (K156TWK) is a specialized immunoassay kit designed for quantitative measurement of interleukin-4 (IL-4) in non-human primate samples This electrochemiluminescence-based assay utilizes Meso Scale Discovery s U-PLEX platform technology offering high sensitivity and specificity for cytokine detection in research applications The kit provides researchers with validated reagents and protocols for studying immune responses and inflammatory processes in preclinical primate models