Learn More
Meso Scale Discovery U-PLEX NHP IL-5 ASY 25 PL
SDP
Catalog No. 5000080631
Encompass
The Meso Scale Discovery U-PLEX NHP IL-5 Assay Kit is a multiplex immunoassay that measures natural human IL-5 levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can be fully automated on the MSD instrument platforms The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrators to generate a standard curve It allows simultaneous measurement of IL-5 alongside other biomarkers in human samples
Catalog No. 50-000-80631 Supplier Meso Scale Discovery Supplier No. K156UOK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-5 Assay Kit is a multiplex immunoassay that measures natural human IL-5 levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision This assay requires 50 L or less sample volume and can be fully automated on the MSD instrument platforms The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrators to generate a standard curve It allows simultaneous measurement of IL-5 alongside other biomarkers in human samples