Learn More
Meso Scale Discovery U-PLEX NHP IL-5 ASY
SDP
Catalog No. 5000080079
Encompass
The Meso Scale Discovery U-PLEX NHP IL-5 Assay Kit is a multiplex immunoassay that measures natural human IL-5 levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Discovery instrumentation including the MESO QuickPlex SQ 120 MESO SECTOR S 600 and MESO SECTOR S 6 Each kit contains a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator
Catalog No. 50-000-80079 Supplier Meso Scale Discovery Supplier No. K156UOK1
The Meso Scale Discovery U-PLEX NHP IL-5 Assay Kit is a multiplex immunoassay that measures natural human IL-5 levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Discovery instrumentation including the MESO QuickPlex SQ 120 MESO SECTOR S 600 and MESO SECTOR S 6 Each kit contains a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator