Learn More
Meso Scale Discovery U-PLEX NHP IL-5 ASY
SDP
Catalog No. 5000080763
Encompass
The Meso Scale Discovery U-PLEX NHP IL-5 Assay Kit is a multiplex immunoassay that measures natural human IL-5 levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Discovery MESO QuickPlex SQ 120 MESO SECTOR S 600 or MESO SECTOR S 6000 instruments The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator It allows the quantitative measurement of natural human IL-5 with a testing time of approximately 3 5 hours
Catalog No. 50-000-80763 Supplier Meso Scale Discovery Supplier No. K156UOK2
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX NHP IL-5 Assay Kit is a multiplex immunoassay that measures natural human IL-5 levels in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption The assay requires 50 L of sample and can be read on Meso Scale Discovery MESO QuickPlex SQ 120 MESO SECTOR S 600 or MESO SECTOR S 6000 instruments The kit includes a plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator It allows the quantitative measurement of natural human IL-5 with a testing time of approximately 3 5 hours