Learn More
Meso Scale Discovery U-PLEX NHP IL-5 ASY384-well
SDP
Catalog No. 5000081233
Encompass
The Meso Scale Discovery U-PLEX NHP IL-5 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring IL-5 in human non-human primate rat and mouse samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive results in 3 5 hours The kit includes a 384-well plate detection antibodies buffers blockers calibrators and controls Requiring only 25 L of sample it is ideal for precious samples with a dynamic range of up to 4 logs quantifying pg/mL to ng/mL levels
Catalog No. 50-000-81233 Supplier Meso Scale Discovery Supplier No. K256UOK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-5 384-Well Assay Kit is an electrochemiluminescent immunoassay for measuring IL-5 in human non-human primate rat and mouse samples It uses MSD s SULFO-TAG labels and multi-array technology for sensitive results in 3 5 hours The kit includes a 384-well plate detection antibodies buffers blockers calibrators and controls Requiring only 25 L of sample it is ideal for precious samples with a dynamic range of up to 4 logs quantifying pg/mL to ng/mL levels