Learn More
Meso Scale Discovery U-PLEX NHP IL-6 ASY
SDP
Catalog No. 5000079966
Encompass
The Meso Scale Discovery U-PLEX NHP IL-6 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 6 (IL-6) in serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a wide dynamic range The assay has high specificity for IL-6 with no cross-reactivity to other cytokines It requires low sample volume and has a rapid procedure time of 3 5 hours The kit includes a 96-well plate coated with IL-6 capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator
Catalog No. 50-000-79966 Supplier Meso Scale Discovery Supplier No. K156TXK2
The Meso Scale Discovery U-PLEX NHP IL-6 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 6 (IL-6) in serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a wide dynamic range The assay has high specificity for IL-6 with no cross-reactivity to other cytokines It requires low sample volume and has a rapid procedure time of 3 5 hours The kit includes a 96-well plate coated with IL-6 capture antibody detection antibody read buffer calibrator diluent wash buffer and a vial of lyophilized calibrator