Learn More
Meso Scale Discovery U-PLEX NHP IL-6 ASY384-well
SDP
Catalog No. 5000080004
Encompass
The Meso Scale Discovery U-PLEX NHP IL-6 384-Well Assay Kit is a multiplex immunoassay that measures human interleukin-6 (IL-6) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrators and controls It allows high-throughput analysis of IL-6 levels with a small sample volume requirement of 25 L per well The kit has a typical sensitivity of 0 06 pg/mL and an upper limit of quantification of 10 000 pg/mL for IL-6
Catalog No. 50-000-80004 Supplier Meso Scale Discovery Supplier No. K256TXK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-6 384-Well Assay Kit is a multiplex immunoassay that measures human interleukin-6 (IL-6) in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrators and controls It allows high-throughput analysis of IL-6 levels with a small sample volume requirement of 25 L per well The kit has a typical sensitivity of 0 06 pg/mL and an upper limit of quantification of 10 000 pg/mL for IL-6