Learn More
Meso Scale Discovery U-PLEX NHP IL-7 ASY 25 PL
SDP
Catalog No. 5000080240
Encompass
The Meso Scale Discovery U-PLEX NHP IL-7 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 0 10 pg/mL and the assay range is 0 10-1000 pg/mL
Catalog No. 50-000-80240 Supplier Meso Scale Discovery Supplier No. K156UPK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-7 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 0 10 pg/mL and the assay range is 0 10-1000 pg/mL