Learn More
Meso Scale Discovery U-PLEX NHP IL-7 ASY
SDP
Catalog No. 5000080632
Encompass
The Meso Scale Discovery U-PLEX NHP IL-7 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 0 3 pg/mL and the assay range is 0 3-2500 pg/mL
Catalog No. 50-000-80632 Supplier Meso Scale Discovery Supplier No. K156UPK1
The Meso Scale Discovery U-PLEX NHP IL-7 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 7 (IL-7) in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The lower limit of detection is 0 3 pg/mL and the assay range is 0 3-2500 pg/mL