Learn More
Meso Scale Discovery U-PLEX NHP IL-7 ASYQP 25 PL
SDP
Catalog No. 5000080078
Encompass
The U-PLEX NHP IL-7 Assay (K156UPK) is a specialized immunoassay kit designed for measuring interleukin-7 levels in non-human primate samples This research tool utilizes Meso Scale Discovery s electrochemiluminescence detection technology to provide sensitive and accurate quantification of IL-7 a crucial cytokine involved in immune system regulation and T-cell development The assay is optimized for serum plasma and cell culture supernatant samples from non-human primates making it valuable for preclinical research and drug development studies
Catalog No. 50-000-80078 Supplier Meso Scale Discovery Supplier No. K156UPK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP IL-7 Assay (K156UPK) is a specialized immunoassay kit designed for measuring interleukin-7 levels in non-human primate samples This research tool utilizes Meso Scale Discovery s electrochemiluminescence detection technology to provide sensitive and accurate quantification of IL-7 a crucial cytokine involved in immune system regulation and T-cell development The assay is optimized for serum plasma and cell culture supernatant samples from non-human primates making it valuable for preclinical research and drug development studies