Learn More
Meso Scale Discovery U-PLEX NHP IL-8 ASY 25 PL
SDP
Catalog No. 5000078551
Encompass
The Meso Scale Discovery U-PLEX NHP IL-8 Assay is a multiplex immunoassay kit for measuring human interleukin 8 (IL-8) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive IL-8 quantification with minimal sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls buffers plate seals and instructions Results are available in 3 5 hours using MSD s quick plex plate reader aiding biomarker and drug discovery research
Catalog No. 50-000-78551 Supplier Meso Scale Discovery Supplier No. K156TYK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-8 Assay is a multiplex immunoassay kit for measuring human interleukin 8 (IL-8) in serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive IL-8 quantification with minimal sample volume The kit includes a 96-well plate with capture antibodies detection antibodies with ruthenium labels calibrators controls buffers plate seals and instructions Results are available in 3 5 hours using MSD s quick plex plate reader aiding biomarker and drug discovery research