Learn More
Meso Scale Discovery U-PLEX NHP IL-8 ASY
SDP
Catalog No. 5000079018
Encompass
The Meso Scale Discovery U-PLEX NHP IL-8 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 8 (IL-8) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 04 pg/mL This assay allows for the accurate and reproducible measurement of IL-8 across a wide range of concentrations The U-PLEX format provides rapid results in just 3 5 hours
Catalog No. 50-000-79018 Supplier Meso Scale Discovery Supplier No. K156TYK2
The Meso Scale Discovery U-PLEX NHP IL-8 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human interleukin 8 (IL-8) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SECTOR Imager and provides high sensitivity a broad dynamic range and requires low sample volume The lower limit of detection is 0 04 pg/mL This assay allows for the accurate and reproducible measurement of IL-8 across a wide range of concentrations The U-PLEX format provides rapid results in just 3 5 hours