Learn More
Meso Scale Discovery U-PLEX NHP IL-8 ASY
SDP
Catalog No. 5000080847
Encompass
The Meso Scale Discovery U-PLEX NHP IL-8 Assay Kit is a sensitive immunoassay for measuring human interleukin 8 (IL-8) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology and works with the MESO QuickPlex SQ 120 instrument The detection range is 0 06-1500 pg/mL with minimal cross-reactivity Each kit assays 40 samples in duplicate on a 96-well plate takes about 3 hours and requires 50 L of sample It offers a rapid ultra-sensitive and cost-effective method for quantifying IL-8 levels
Catalog No. 50-000-80847 Supplier Meso Scale Discovery Supplier No. K156TYK1
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX NHP IL-8 Assay Kit is a sensitive immunoassay for measuring human interleukin 8 (IL-8) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology and works with the MESO QuickPlex SQ 120 instrument The detection range is 0 06-1500 pg/mL with minimal cross-reactivity Each kit assays 40 samples in duplicate on a 96-well plate takes about 3 hours and requires 50 L of sample It offers a rapid ultra-sensitive and cost-effective method for quantifying IL-8 levels