Learn More
Meso Scale Discovery U-PLEX NHP IL-9 ASY 25 PL
SDP
Catalog No. 5000080738
Encompass
The Meso Scale Discovery U-PLEX NHP IL-9 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human IL-9 in serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a wide dynamic range The assay has high specificity and no cross-reactivity It requires low sample volume and has a rapid procedure time of 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Blend and Sample Diluent It has a sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL
Catalog No. 50-000-80738 Supplier Meso Scale Discovery Supplier No. K156XKK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IL-9 Assay is an electrochemiluminescent immunoassay for the quantitative measurement of human IL-9 in serum plasma and cell culture supernatants It utilizes MSD s proprietary SECTOR Imager 6000 for high sensitivity and a wide dynamic range The assay has high specificity and no cross-reactivity It requires low sample volume and has a rapid procedure time of 3 5 hours The kit includes a 96-well plate coated with capture antibody detection antibody Read Buffer Diluent 2 Calibrator Blend and Sample Diluent It has a sensitivity of 0 04 pg/mL and an upper limit of quantification of 2500 pg/mL