Learn More
Meso Scale Discovery U-PLEX NHP IL-9 ASY
SDP
Catalog No. 5000078495
Encompass
The Meso Scale Discovery U-PLEX NHP IL-9 Assay Kit is a multiplex immunoassay that enables simultaneous quantification of IL-9 biomarker levels in non-human primate (NHP) samples such as serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements It includes a 96-well plate pre-coated with capture antibodies for IL-9 calibrators detection antibodies read buffer wash buffer and plate seals The kit can test up to 80 samples in duplicates and has a testing time of 3 5 hours
Catalog No. 50-000-78495 Supplier Meso Scale Discovery Supplier No. K156XKK2
The Meso Scale Discovery U-PLEX NHP IL-9 Assay Kit is a multiplex immunoassay that enables simultaneous quantification of IL-9 biomarker levels in non-human primate (NHP) samples such as serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements It includes a 96-well plate pre-coated with capture antibodies for IL-9 calibrators detection antibodies read buffer wash buffer and plate seals The kit can test up to 80 samples in duplicates and has a testing time of 3 5 hours