Learn More
Meso Scale Discovery U-PLEX NHP IL-9 ASY
SDP
Catalog No. 5000081067
Encompass
The Meso Scale Discovery U-PLEX NHP IL-9 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human IL-9 in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The kit has a standard curve range from 0 61-2500 pg/mL and exhibits no cross-reactivity with other cytokines The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-81067 Supplier Meso Scale Discovery Supplier No. K156XKK1
The Meso Scale Discovery U-PLEX NHP IL-9 Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of human IL-9 in serum plasma and cell culture supernatants The assay uses MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results with low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies conjugated to ruthenium labels calibrators and reagents required to perform the assay The kit has a standard curve range from 0 61-2500 pg/mL and exhibits no cross-reactivity with other cytokines The assay takes approximately 3 5 hours to complete