Learn More
Meso Scale Discovery U-PLEX NHP IP-10 ASY 25 PL
SDP
Catalog No. 5000079069
Encompass
The Meso Scale Discovery U-PLEX NHP IP-10 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human interferon gamma-induced protein 10 (IP-10) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The assay has high sensitivity and wide dynamic range and requires only 25 L sample volume per well The total assay time is approximately 3 5 hours This product can aid in understanding IP-10 biology and evaluating IP-10 as a biomarker across disease areas including autoimmune disorders infectious diseases and oncology
Catalog No. 50-000-79069 Supplier Meso Scale Discovery Supplier No. K156UFK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP IP-10 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human interferon gamma-induced protein 10 (IP-10) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The assay has high sensitivity and wide dynamic range and requires only 25 L sample volume per well The total assay time is approximately 3 5 hours This product can aid in understanding IP-10 biology and evaluating IP-10 as a biomarker across disease areas including autoimmune disorders infectious diseases and oncology