Learn More
Meso Scale Discovery U-PLEX NHP IP-10 ASY
SDP
Catalog No. 5000080854
Encompass
The U-PLEX NHP IP-10 Assay is a multiplex immunoassay panel that measures 10 biomarkers including IP-10 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology on the Meso Scale Discovery platform The assay has high sensitivity broad dynamic range and requires low sample volume It is designed for research use only to study immune system disorders infections and inflammatory diseases The kit includes a 10-plex plate diluent calibrators and controls to run the immunoassay
Catalog No. 50-000-80854 Supplier Meso Scale Discovery Supplier No. K156UFK1
The U-PLEX NHP IP-10 Assay is a multiplex immunoassay panel that measures 10 biomarkers including IP-10 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology on the Meso Scale Discovery platform The assay has high sensitivity broad dynamic range and requires low sample volume It is designed for research use only to study immune system disorders infections and inflammatory diseases The kit includes a 10-plex plate diluent calibrators and controls to run the immunoassay