Learn More
Meso Scale Discovery U-PLEX NHP IP-10 ASYQP 25 PL
SDP
Catalog No. 5000079950
Encompass
The K156UFK-2 is a multiplex immunoassay kit from Meso Scale Discovery that enables simultaneous detection and quantification of multiple biomarkers in a single sample This electrochemiluminescence-based assay platform provides high sensitivity wide dynamic range and reduced sample volume requirements compared to traditional ELISA methods The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for streamlined workflow Designed for research applications it offers superior precision and reproducibility for biomarker analysis in various sample types including serum plasma and cell culture supernatants
Catalog No. 50-000-79950 Supplier Meso Scale Discovery Supplier No. K156UFK24
Add to Cart
Add to Cart
Request a Quote
The K156UFK-2 is a multiplex immunoassay kit from Meso Scale Discovery that enables simultaneous detection and quantification of multiple biomarkers in a single sample This electrochemiluminescence-based assay platform provides high sensitivity wide dynamic range and reduced sample volume requirements compared to traditional ELISA methods The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for streamlined workflow Designed for research applications it offers superior precision and reproducibility for biomarker analysis in various sample types including serum plasma and cell culture supernatants