Learn More
Meso Scale Discovery U-PLEX NHP IP-10 ASYQP
SDP
Catalog No. 5000078559
Encompass
MSD s U-PLEX IP-10 Singleplex Assay is designed for non-human primate samples utilizing electrochemiluminescence detection technology for precise cytokine measurement This research-grade immunoassay provides reliable quantification of IP-10 (CXCL10) a key chemokine involved in immune responses and inflammation studies The U-PLEX platform offers superior sensitivity and dynamic range compared to traditional ELISA methods making it ideal for preclinical research applications requiring accurate biomarker analysis in primate models
Catalog No. 50-000-78559 Supplier Meso Scale Discovery Supplier No. K156UFK21
MSD s U-PLEX IP-10 Singleplex Assay is designed for non-human primate samples utilizing electrochemiluminescence detection technology for precise cytokine measurement This research-grade immunoassay provides reliable quantification of IP-10 (CXCL10) a key chemokine involved in immune responses and inflammation studies The U-PLEX platform offers superior sensitivity and dynamic range compared to traditional ELISA methods making it ideal for preclinical research applications requiring accurate biomarker analysis in primate models