Learn More
Meso Scale Discovery U-PLEX NHP M-CSF ASY 25 PL
SDP
Catalog No. 5000080899
Encompass
The U-PLEX NHP CSF Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that measures neurofilament light chain (NfL) total tau (t-tau) phosphorylated tau (p-tau181) amyloid beta 42 (A 42) amyloid beta 40 (A 40) YKL-40 neurogranin and sTREM2 in cerebrospinal fluid (CSF) This 8-plex assay utilizes electrochemiluminescence (ECL) detection technology and provides quantitative results in under 3 hours from a small CSF sample volume It is intended for investigational use by biopharmaceutical companies and clinical researchers to aid in understanding neurodegenerative diseases like Alzheimer s
Catalog No. 50-000-80899 Supplier Meso Scale Discovery Supplier No. K156XRK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP CSF Assay from Meso Scale Diagnostics is a multiplex immunoassay panel that measures neurofilament light chain (NfL) total tau (t-tau) phosphorylated tau (p-tau181) amyloid beta 42 (A 42) amyloid beta 40 (A 40) YKL-40 neurogranin and sTREM2 in cerebrospinal fluid (CSF) This 8-plex assay utilizes electrochemiluminescence (ECL) detection technology and provides quantitative results in under 3 hours from a small CSF sample volume It is intended for investigational use by biopharmaceutical companies and clinical researchers to aid in understanding neurodegenerative diseases like Alzheimer s