Learn More
Meso Scale Discovery U-PLEX NHP M-CSF ASY
SDP
Catalog No. 5000078490
Encompass
The Meso Scale Discovery U-PLEX NHP CSF Assay is a multiplex immunoassay that measures NfL t-tau p-tau A 42 A 40 YKL-40 and neurogranin in human CSF It uses ECL detection technology providing quantitative results in under 5 hours from a small sample Intended for investigational use by pharmaceutical companies and researchers it aids in understanding neurodegenerative diseases like Alzheimer s The kit includes a 96-well plate detection antibodies calibrators controls assay diluent read buffer wash buffer and plate seals
Catalog No. 50-000-78490 Supplier Meso Scale Discovery Supplier No. K156XRK1
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX NHP CSF Assay is a multiplex immunoassay that measures NfL t-tau p-tau A 42 A 40 YKL-40 and neurogranin in human CSF It uses ECL detection technology providing quantitative results in under 5 hours from a small sample Intended for investigational use by pharmaceutical companies and researchers it aids in understanding neurodegenerative diseases like Alzheimer s The kit includes a 96-well plate detection antibodies calibrators controls assay diluent read buffer wash buffer and plate seals