Learn More
Meso Scale Discovery U-PLEX NHP M-CSF ASY384-well
SDP
Catalog No. 5000080271
Encompass
The U-PLEX NHP M CSF 384-Well Assay from Meso Scale Diagnostics is a 384-well multiplex immunoassay panel that measures 38 biomarkers related to neuroinflammation and neurodegeneration in human cerebrospinal fluid (CSF) It utilizes electrochemiluminescence detection for high sensitivity and broad dynamic range This assay allows researchers to profile biomarkers associated with various neurological conditions efficiently from low CSF volumes It provides a streamlined workflow using premixed lyophilized reagents for minimal hands-on time The panel includes analytes such as A peptides tau proteins -synuclein NFL GFAP sTREM2 YKL-40 and more
Catalog No. 50-000-80271 Supplier Meso Scale Discovery Supplier No. K256XRK2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP M CSF 384-Well Assay from Meso Scale Diagnostics is a 384-well multiplex immunoassay panel that measures 38 biomarkers related to neuroinflammation and neurodegeneration in human cerebrospinal fluid (CSF) It utilizes electrochemiluminescence detection for high sensitivity and broad dynamic range This assay allows researchers to profile biomarkers associated with various neurological conditions efficiently from low CSF volumes It provides a streamlined workflow using premixed lyophilized reagents for minimal hands-on time The panel includes analytes such as A peptides tau proteins -synuclein NFL GFAP sTREM2 YKL-40 and more