Learn More
Meso Scale Discovery U-PLEX NHP M-CSF ASYQP
SDP
Catalog No. 5000080898
Encompass
The U-PLEX NHP M-CSF Assay (K156XRK) is a multiplex immunoassay designed to measure Macrophage Colony-Stimulating Factor (M-CSF) levels in non-human primate samples This research tool utilizes Meso Scale Discovery s electrochemiluminescence detection technology for sensitive and accurate quantification of M-CSF biomarkers in biological samples supporting preclinical research applications
Catalog No. 50-000-80898 Supplier Meso Scale Discovery Supplier No. K156XRK22
The U-PLEX NHP M-CSF Assay (K156XRK) is a multiplex immunoassay designed to measure Macrophage Colony-Stimulating Factor (M-CSF) levels in non-human primate samples This research tool utilizes Meso Scale Discovery s electrochemiluminescence detection technology for sensitive and accurate quantification of M-CSF biomarkers in biological samples supporting preclinical research applications