Learn More
Meso Scale Discovery U-PLEX NHP MCP-1 ASY 25 PL
SDP
Catalog No. 5000079776
Encompass
The Meso Scale Discovery U-PLEX NHP MCP-1 Assay is a multiplex immunoassay kit for measuring human Monocyte Chemoattractant Protein 1 (MCP-1) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls Turnaround time is 3 5 hours for up to 80 samples aiding in MCP-1 biology understanding and therapy evaluation
Catalog No. 50-000-79776 Supplier Meso Scale Discovery Supplier No. K156UGK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP MCP-1 Assay is a multiplex immunoassay kit for measuring human Monocyte Chemoattractant Protein 1 (MCP-1) in serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls Turnaround time is 3 5 hours for up to 80 samples aiding in MCP-1 biology understanding and therapy evaluation