Learn More
Meso Scale Discovery U-PLEX NHP MCP-1 ASY
SDP
Catalog No. 5000079951
Encompass
The Meso Scale Discovery U-PLEX NHP MCP-1 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human Monocyte Chemoattractant Protein 1 (MCP-1) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This assay allows rapid and reliable quantification of MCP-1 levels in human samples
Catalog No. 50-000-79951 Supplier Meso Scale Discovery Supplier No. K156UGK1
The Meso Scale Discovery U-PLEX NHP MCP-1 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human Monocyte Chemoattractant Protein 1 (MCP-1) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and validate assay performance This assay allows rapid and reliable quantification of MCP-1 levels in human samples