Learn More
Meso Scale Discovery U-PLEX NHP MCP-1 ASY
SDP
Catalog No. 5000081147
Encompass
The Meso Scale Discovery U-PLEX NHP MCP-1 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human Monocyte Chemoattractant Protein 1 (MCP-1) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions for generating a standard curve This assay provides high sensitivity a wide dynamic range and requires a low sample volume of 50 L per well
Catalog No. 50-000-81147 Supplier Meso Scale Discovery Supplier No. K156UGK2
The Meso Scale Discovery U-PLEX NHP MCP-1 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of human Monocyte Chemoattractant Protein 1 (MCP-1) in serum plasma and cell culture supernatants This assay utilizes MSD s proprietary electrochemiluminescent detection technology and is provided in a convenient 96-well plate format The kit includes a plate coated with capture antibodies detection antibodies labeled with an electrochemiluminescent compound and calibrator solutions for generating a standard curve This assay provides high sensitivity a wide dynamic range and requires a low sample volume of 50 L per well