Learn More
Meso Scale Discovery U-PLEX NHP MCP-2 ASY 25 PL
SDP
Catalog No. 5000080234
Encompass
The U-PLEX NHP MCP-2 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of NHP MCP-2 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay requires an MSD instrument and takes approximately 3 5 hours to complete with only a few pipetting steps required by the user It includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator The kit can test up to 80 samples in duplicate
Catalog No. 50-000-80234 Supplier Meso Scale Discovery Supplier No. K156XHK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP MCP-2 Assay is a multiplex immunoassay kit from Meso Scale Diagnostics that allows simultaneous quantitative measurement of NHP MCP-2 in serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The assay requires an MSD instrument and takes approximately 3 5 hours to complete with only a few pipetting steps required by the user It includes a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 and calibrator The kit can test up to 80 samples in duplicate