Learn More
Meso Scale Discovery U-PLEX NHP MCP-2 ASY
SDP
Catalog No. 5000080735
Encompass
The U-PLEX NHP MCP-2 Assay by Meso Scale Diagnostics is a multiplex immunoassay kit for measuring NHP MCP-2 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity broad dynamic range and low sample volume The assay requires an MSD instrument takes about 3 5 hours and involves minimal pipetting It includes a 96-well plate with capture antibodies detection antibodies buffers diluent and calibrators The kit is highly specific with under 0 3% cross-reactivity and is for research use only
Catalog No. 50-000-80735 Supplier Meso Scale Discovery Supplier No. K156XHK2
The U-PLEX NHP MCP-2 Assay by Meso Scale Diagnostics is a multiplex immunoassay kit for measuring NHP MCP-2 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection offering high sensitivity broad dynamic range and low sample volume The assay requires an MSD instrument takes about 3 5 hours and involves minimal pipetting It includes a 96-well plate with capture antibodies detection antibodies buffers diluent and calibrators The kit is highly specific with under 0 3% cross-reactivity and is for research use only