Learn More
Meso Scale Discovery U-PLEX NHP MCP-2 ASY384-well
SDP
Catalog No. 5000078458
Encompass
The U-PLEX NHP MCP-2 (384-well assay) K256XHK is a 384-well assay kit for measuring human MCP-2 in serum plasma urine cell culture supernatants and other biological fluids It uses electrochemiluminescence (ECL) technology and provides accurate reproducible results in just 3 5 hours The kit includes a 384-well MCP-2 plate detection antibody and all necessary reagents and buffers It has a wide dynamic range of 78-50 000 pg/mL and requires just 25 L sample size per well This assay kit offers high sensitivity specificity and lot-to-lot consistency for reliable MCP-2 quantification
Catalog No. 50-000-78458 Supplier Meso Scale Discovery Supplier No. K256XHK2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP MCP-2 (384-well assay) K256XHK is a 384-well assay kit for measuring human MCP-2 in serum plasma urine cell culture supernatants and other biological fluids It uses electrochemiluminescence (ECL) technology and provides accurate reproducible results in just 3 5 hours The kit includes a 384-well MCP-2 plate detection antibody and all necessary reagents and buffers It has a wide dynamic range of 78-50 000 pg/mL and requires just 25 L sample size per well This assay kit offers high sensitivity specificity and lot-to-lot consistency for reliable MCP-2 quantification