Learn More
Meso Scale Discovery U-PLEX NHP MCP-2 ASYQP 25 PL
SDP
Catalog No. 5000078496
Encompass
MSD s U-PLEX MCP-2 Singleplex Assay for non-human primates is an electrochemiluminescence-based immunoassay designed to quantify monocyte chemoattractant protein-2 levels in primate samples This research tool offers high sensitivity and specificity for measuring MCP-2 a key inflammatory biomarker involved in immune cell recruitment and inflammatory responses The U-PLEX platform provides reliable reproducible results for preclinical research applications studying inflammation immunology and disease pathways in non-human primate models
Catalog No. 50-000-78496 Supplier Meso Scale Discovery Supplier No. K156XHK24
Add to Cart
Add to Cart
Request a Quote
MSD s U-PLEX MCP-2 Singleplex Assay for non-human primates is an electrochemiluminescence-based immunoassay designed to quantify monocyte chemoattractant protein-2 levels in primate samples This research tool offers high sensitivity and specificity for measuring MCP-2 a key inflammatory biomarker involved in immune cell recruitment and inflammatory responses The U-PLEX platform provides reliable reproducible results for preclinical research applications studying inflammation immunology and disease pathways in non-human primate models