Learn More
Meso Scale Discovery U-PLEX NHP MDC ASY 25 PL
SDP
Catalog No. 5000080056
Encompass
The U-PLEX NHP MDC Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay panel that enables simultaneous quantitative measurement of up to 10 biomarkers in human serum plasma or cell culture supernatants This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals This assay allows efficient profiling of biomarkers associated with neuroinflammation and neurodegeneration
Catalog No. 50-000-80056 Supplier Meso Scale Discovery Supplier No. K156UIK4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP MDC Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay panel that enables simultaneous quantitative measurement of up to 10 biomarkers in human serum plasma or cell culture supernatants This assay utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and requires low sample volume The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals This assay allows efficient profiling of biomarkers associated with neuroinflammation and neurodegeneration