Learn More
Meso Scale Discovery U-PLEX NHP MDC ASY
SDP
Catalog No. 5000080057
Encompass
The U-PLEX NHP MDC Assay K156UIK from Meso Scale Diagnostics is a multiplex immunoassay kit that enables simultaneous quantitative measurement of up to 10 biomarkers in human serum plasma and cell culture supernatants This assay uses electrochemiluminescent detection technology for high sensitivity and broad dynamic range The kit includes a 10-plex human panel containing IFN-gamma IL-10 IL-12p70 IL-13 IL-1beta IL-2 IL-4 IL-6 TNF-alpha and GM-CSF It requires 50 L or less sample volume and can perform up to 80 tests across two plates The kit includes all necessary reagents and plates for the assay
Catalog No. 50-000-80057 Supplier Meso Scale Discovery Supplier No. K156UIK1
Add to Cart
Add to Cart
The U-PLEX NHP MDC Assay K156UIK from Meso Scale Diagnostics is a multiplex immunoassay kit that enables simultaneous quantitative measurement of up to 10 biomarkers in human serum plasma and cell culture supernatants This assay uses electrochemiluminescent detection technology for high sensitivity and broad dynamic range The kit includes a 10-plex human panel containing IFN-gamma IL-10 IL-12p70 IL-13 IL-1beta IL-2 IL-4 IL-6 TNF-alpha and GM-CSF It requires 50 L or less sample volume and can perform up to 80 tests across two plates The kit includes all necessary reagents and plates for the assay