Learn More
Meso Scale Discovery U-PLEX NHP MDC ASY
SDP
Catalog No. 5000080643
Encompass
The U-PLEX NHP MDC Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for detecting up to 10 biomarkers in human serum plasma and cell culture supernatants It uses electrochemiluminescent detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals It quantifies biomarkers related to neurodegenerative diseases and neuroinflammation including Abeta tau alpha-synuclein NFL GFAP sTREM2 YKL-40 IL-6 IL-8 and TNF-alpha
Catalog No. 50-000-80643 Supplier Meso Scale Discovery Supplier No. K156UIK2
The U-PLEX NHP MDC Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay for detecting up to 10 biomarkers in human serum plasma and cell culture supernatants It uses electrochemiluminescent detection for high sensitivity and broad dynamic range with low sample volume The kit includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals It quantifies biomarkers related to neurodegenerative diseases and neuroinflammation including Abeta tau alpha-synuclein NFL GFAP sTREM2 YKL-40 IL-6 IL-8 and TNF-alpha