Learn More
Meso Scale Discovery U-PLEX NHP MDC ASY384-well
SDP
Catalog No. 5000081228
Encompass
The U-PLEX NHP MDC Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that enables simultaneous quantitative measurement of up to 10 biomarkers in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) assay utilizes MSD s proprietary U-PLEX technology providing high sensitivity a large dynamic range and minimal matrix interference The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls Biomarkers measured include IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 IFN- TNF- GM-CSF and IL-1 The kit is for research use only
Catalog No. 50-000-81228 Supplier Meso Scale Discovery Supplier No. K256UIK2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP MDC Assay Kit from Meso Scale Diagnostics is a multiplex immunoassay that enables simultaneous quantitative measurement of up to 10 biomarkers in human serum plasma and cell culture supernatants This electrochemiluminescence (ECL) assay utilizes MSD s proprietary U-PLEX technology providing high sensitivity a large dynamic range and minimal matrix interference The kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators and controls Biomarkers measured include IL-2 IL-4 IL-5 IL-10 IL-12p70 IL-13 IFN- TNF- GM-CSF and IL-1 The kit is for research use only