Learn More
Meso Scale Discovery U-PLEX NHP MIF ASY 25 PL
SDP
Catalog No. 5000081066
Encompass
The Meso Scale Discovery U-PLEX NHP MIF Assay is a multiplex immunoassay kit for quantifying macrophage migration inhibitory factor (MIF) in non-human primate samples like serum and plasma It uses MSD s electrochemiluminescent detection on the QuickPlex SQ 120 instrument measuring MIF from 50-10 000 pg/mL with minimal sample volume The kit includes a plate with capture antibodies detection antibodies calibrators controls diluent read buffer and necessary reagents It offers high sensitivity a wide dynamic range and requires 50 L or less of sample
Catalog No. 50-000-81066 Supplier Meso Scale Discovery Supplier No. K156XJK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP MIF Assay is a multiplex immunoassay kit for quantifying macrophage migration inhibitory factor (MIF) in non-human primate samples like serum and plasma It uses MSD s electrochemiluminescent detection on the QuickPlex SQ 120 instrument measuring MIF from 50-10 000 pg/mL with minimal sample volume The kit includes a plate with capture antibodies detection antibodies calibrators controls diluent read buffer and necessary reagents It offers high sensitivity a wide dynamic range and requires 50 L or less of sample