Learn More
Meso Scale Discovery U-PLEX NHP MIF ASY
SDP
Catalog No. 5000080913
Encompass
The Meso Scale Discovery U-PLEX NHP MIF Assay is a multiplex immunoassay kit for measuring macrophage migration inhibitory factor (MIF) in non-human primate samples like serum and plasma It uses MSD s electrochemiluminescent detection in a 96-well plate format The kit includes capture and detection antibodies calibrators controls diluent read buffer and necessary reagents It has a sensitivity of 0 04 pg/mL and a range of 0 08 to 400 pg/mL offering superior sensitivity and quantitation compared to conventional ELISA assays
Catalog No. 50-000-80913 Supplier Meso Scale Discovery Supplier No. K156XJK1
Add to Cart
Add to Cart
The Meso Scale Discovery U-PLEX NHP MIF Assay is a multiplex immunoassay kit for measuring macrophage migration inhibitory factor (MIF) in non-human primate samples like serum and plasma It uses MSD s electrochemiluminescent detection in a 96-well plate format The kit includes capture and detection antibodies calibrators controls diluent read buffer and necessary reagents It has a sensitivity of 0 04 pg/mL and a range of 0 08 to 400 pg/mL offering superior sensitivity and quantitation compared to conventional ELISA assays