Learn More
Meso Scale Discovery U-PLEX NHP MIF ASY
SDP
Catalog No. 5000080914
Encompass
The Meso Scale Discovery U-PLEX NHP MIF Assay is a multiplex immunoassay kit for detecting and quantifying Macrophage Migration Inhibitory Factor (MIF) in non-human primate samples like serum and plasma It uses MSD s electrochemiluminescent technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture and detection antibodies plus calibrators and controls for standard curve generation and assay validation enabling efficient analysis of MIF a key biomarker in NHP studies
Catalog No. 50-000-80914 Supplier Meso Scale Discovery Supplier No. K156XJK2
The Meso Scale Discovery U-PLEX NHP MIF Assay is a multiplex immunoassay kit for detecting and quantifying Macrophage Migration Inhibitory Factor (MIF) in non-human primate samples like serum and plasma It uses MSD s electrochemiluminescent technology offering high sensitivity a wide dynamic range and low sample volume needs The kit includes a 96-well plate with capture and detection antibodies plus calibrators and controls for standard curve generation and assay validation enabling efficient analysis of MIF a key biomarker in NHP studies