Learn More
Meso Scale Discovery U-PLEX NHP MIF ASY384-well
SDP
Catalog No. 5000081755
Encompass
The U-PLEX NHP MIF 384-Well Assay from Meso Scale Discovery is a 384-well multiplex immunoassay that enables simultaneous detection of multiple human neurodegenerative biomarkers This assay measures A 38 A 40 A 42 -synuclein p-tau181 t-tau and neurogranin in human CSF and plasma samples using electrochemiluminescence detection technology The assay utilizes Meso Scale Discovery s proprietary U-PLEX technology and provides high sensitivity broad dynamic range and minimal matrix interference for biomarker quantification With a small sample volume requirement and 384-well format this assay allows efficient processing of large sample sets
Catalog No. 50-000-81755 Supplier Meso Scale Discovery Supplier No. K256XJK2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX NHP MIF 384-Well Assay from Meso Scale Discovery is a 384-well multiplex immunoassay that enables simultaneous detection of multiple human neurodegenerative biomarkers This assay measures A 38 A 40 A 42 -synuclein p-tau181 t-tau and neurogranin in human CSF and plasma samples using electrochemiluminescence detection technology The assay utilizes Meso Scale Discovery s proprietary U-PLEX technology and provides high sensitivity broad dynamic range and minimal matrix interference for biomarker quantification With a small sample volume requirement and 384-well format this assay allows efficient processing of large sample sets