Learn More
Meso Scale Discovery U-PLEX NHP MIP-1 ASY 25 PL
SDP
Catalog No. 5000080050
Encompass
The Meso Scale Discovery U-PLEX NHP MIP-1alpha Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of mouse and rat MIP-1alpha in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on MSD s SECTOR Imager 6000 or QuickPlex SQ 120 instruments The kit includes a plate coated with capture antibodies detection antibodies and calibrators ranging from 500 to 0 61 pg/mL
Catalog No. 50-000-80050 Supplier Meso Scale Discovery Supplier No. K156UJK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX NHP MIP-1alpha Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of mouse and rat MIP-1alpha in serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on MSD s SECTOR Imager 6000 or QuickPlex SQ 120 instruments The kit includes a plate coated with capture antibodies detection antibodies and calibrators ranging from 500 to 0 61 pg/mL