Learn More
Meso Scale Discovery U-PLEX NHP MIP-1 ASY
SDP
Catalog No. 5000080642
Encompass
The Meso Scale Discovery U-PLEX NHP MIP-1alpha Assay Kit is a multiplex immunoassay that measures human macrophage inflammatory protein 1-alpha (MIP-1alpha) in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to quantify MIP-1alpha concentrations
Catalog No. 50-000-80642 Supplier Meso Scale Discovery Supplier No. K156UJK1
The Meso Scale Discovery U-PLEX NHP MIP-1alpha Assay Kit is a multiplex immunoassay that measures human macrophage inflammatory protein 1-alpha (MIP-1alpha) in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on MSD instrument platforms It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to quantify MIP-1alpha concentrations